Learn More
CAS DATALOGGERS dataTaker DT80 Universal Data Logger
Catalog No. NC3846995
Encompass
"The dataTaker DT80 Universal Input Data Logger provides an extensive array of features that allow it to be used across various applications. The DT80 is a robust, stand-alone, low-power data logger featuring USB memory stick support, 18-bit resolution, extensive communications capabilities, and a built-in display.The dataTaker DT80’s Dual Channel concept allows up to 10 isolated or 15 commonly referenced analog inputs to be used in many combinations. With support for multiple SDI-12 sensor networks, Modbus for SCADA systems, FTP and Web interface, and 12V regulated output to power sensors, the DT80 is a self-contained solution."
Catalog No. NC3846995 Supplier CAS DATALOGGERS Supplier No. DT80
"The dataTaker DT80 Universal Input Data Logger provides an extensive array of features that allow it to be used across various applications. The DT80 is a robust, stand-alone, low-power data logger featuring USB memory stick support, 18-bit resolution, extensive communications capabilities, and a built-in display.The dataTaker DT80’s Dual Channel concept allows up to 10 isolated or 15 commonly referenced analog inputs to be used in many combinations. With support for multiple SDI-12 sensor networks, Modbus for SCADA systems, FTP and Web interface, and 12V regulated output to power sensors, the DT80 is a self-contained solution."