Learn More
CAS DATALOGGERS dataTaker DT85 Universal Data Logger
Catalog No. NC3846998
Encompass
"The dataTaker DT85 smart data logger provides an extensive array of features that allow it to be used across a wide variety of applications. The DT85 is a robust, standalone, low power data logger featuring USB memory stick support, 18-bit resolution, extensive communications capabilities and built-in display.The dataTaker DT85’s Dual Channel concept allows up to 32 isolated or 48 common referenced analog inputs to be used in many combinations. With support for multiple SDI-12 sensor networks, Modbus for SCADA systems, FTP and Web interface, 12V regulated output to power sensors, the DT85 is a totally self-contained solution."
Catalog No. NC3846998 Supplier CAS DATALOGGERS Supplier No. DT85
"The dataTaker DT85 smart data logger provides an extensive array of features that allow it to be used across a wide variety of applications. The DT85 is a robust, standalone, low power data logger featuring USB memory stick support, 18-bit resolution, extensive communications capabilities and built-in display.The dataTaker DT85’s Dual Channel concept allows up to 32 isolated or 48 common referenced analog inputs to be used in many combinations. With support for multiple SDI-12 sensor networks, Modbus for SCADA systems, FTP and Web interface, 12V regulated output to power sensors, the DT85 is a totally self-contained solution."