Learn More
Alkali Scientific Date Rec'd/Date Opened/Exp Date Labels, Reagent / Analysis / Date Init.
SDP
Catalog No. 503026742
Encompass_Preferred
Date Rec'd/Date Opened/Exp Date Labels We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).
Catalog No. 50-302-6742 Supplier Alkali Scientific Supplier No. LREA2
Date Rec'd/Date Opened/Exp Date Labels We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).