Learn More
Medchemexpress LLC Datelliptium chloride | 105118-14-7 | 397.94 | 1 ML
SDP
Catalog No. 5000448363
Encompass_Preferred
Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, with anti-tumor activities.
  • DNA-intercalating agent
  • Derived from ellipticine
  • Exhibits anti-tumor activities
Catalog No. 50-004-48363 Supplier Medchemexpress LLC Supplier No. HYU0033710MM/1ML
Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, with anti-tumor activities.
  • DNA-intercalating agent
  • Derived from ellipticine
  • Exhibits anti-tumor activities