Learn More
Medchemexpress LLC Datelliptium (chloride) | 105118-14-7 | 397.94 | 25 MG
SDP
Catalog No. 5000449814
Encompass_Preferred
Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, known for its anti-tumor activities. It demonstrates significant cytotoxic effects in various cell systems.
  • DNA-intercalating agent
  • Exhibits anti-tumor activities
  • Demonstrates significant cytotoxic effects
  • Mobilizes glycogen in rat hepatocytes
  • Decreases gluconeogenesis from lactate
  • Selectively cytotoxic for solid tumors
Catalog No. 50-004-49814 Supplier Medchemexpress LLC Supplier No. 25MGHYU00337
Add to Cart
Add to Cart
Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, known for its anti-tumor activities. It demonstrates significant cytotoxic effects in various cell systems.
  • DNA-intercalating agent
  • Exhibits anti-tumor activities
  • Demonstrates significant cytotoxic effects
  • Mobilizes glycogen in rat hepatocytes
  • Decreases gluconeogenesis from lactate
  • Selectively cytotoxic for solid tumors