Learn More
Medchemexpress LLC Datelliptium chloride | 105118-14-7 | 397.94 | 5 MG
SDP
Catalog No. 5000448321
Encompass_Preferred
Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, exhibiting anti-tumor activities. It demonstrates significant cytotoxic effects in suspension and primary cultures of rat hepatocytes, mobilizing glycogen in vitro in a concentration-dependent manner. This agent also decreases gluconeogenesis from lactate in cells and has shown potent inhibitory activities against various murine solid tumors.
  • DNA-intercalating agent
  • Exhibits anti-tumor activities
  • Shows cytotoxic effects in rat hepatocytes
  • Mobilizes glycogen in rat systems in vitro
  • Decreases gluconeogenesis from lactate in cells
  • Selectively cytotoxic for solid tumors over leukemia L1210 (as datelliptium acetate)
  • Potent inhibitory activities against murine solid tumors in vivo (as datelliptium acetate)
Catalog No. 50-004-48321 Supplier Medchemexpress LLC Supplier No. HYU003375MG
Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, exhibiting anti-tumor activities. It demonstrates significant cytotoxic effects in suspension and primary cultures of rat hepatocytes, mobilizing glycogen in vitro in a concentration-dependent manner. This agent also decreases gluconeogenesis from lactate in cells and has shown potent inhibitory activities against various murine solid tumors.
  • DNA-intercalating agent
  • Exhibits anti-tumor activities
  • Shows cytotoxic effects in rat hepatocytes
  • Mobilizes glycogen in rat systems in vitro
  • Decreases gluconeogenesis from lactate in cells
  • Selectively cytotoxic for solid tumors over leukemia L1210 (as datelliptium acetate)
  • Potent inhibitory activities against murine solid tumors in vivo (as datelliptium acetate)