Learn More
TARGETMOL CHEMICALS INC Datelliptium chloride hydrochl
SDP
Catalog No. 502876317
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Datelliptium chloride hydrochloride is a DNA-intercalating agent derived from ellipticine, exhibiting anti-tumor activities. Purity 99.91%
Catalog No. 50-287-6317 Supplier TARGETMOL CHEMICALS INC Supplier No. T77471MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Datelliptium chloride hydrochloride is a DNA-intercalating agent derived from ellipticine, exhibiting anti-tumor activities. Purity 99.91%