Learn More
Medchemexpress LLC Datopotamab deruxtecan | 2238831-60-0 | 99.5% | 149000 (average) | 5 MG

Supplier: Medchemexpress LLC HY1415985MG
Datopotamab deruxtecan is a TROP2-directed antibody-drug conjugate (ADC) that uses DXd linker-payload technology to deliver a deruxtecan-class topoisomerase I inhibitor to TROP2-expressing tumor cells. Regulatory and safety documents list the CAS number 2238831-60-0.
- TROP2-directed antibody-drug conjugate mechanism
- Uses DXd linker-payload technology to deliver deruxtecan class payload
- Molecular weight approximately 149,000 (average)
- Purity 99.5%
- Storage: -80°C, protect from light
- Available in small research pack sizes such as 5 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.