Learn More
New England Biolabs, Inc. dATP Solution – 25 µmol
SDP
Catalog No. 504411
Encompass
Contains 0.25 ml of 100 mM ultrapure dATP. Supplied in: Ultrapure water as a sodium salt at pH 7.4. Concentration: The dATP Solution is supplied at a concentration of 100 mM as determined by A260 absorbance.
Catalog No. 50-441-1 Supplier New England Biolabs, Inc. Supplier No. N0440S
Contains 0.25 ml of 100 mM ultrapure dATP. Supplied in: Ultrapure water as a sodium salt at pH 7.4. Concentration: The dATP Solution is supplied at a concentration of 100 mM as determined by A260 absorbance.