Learn More
Sigma Aldrich Fine Chemicals Biosciences dATP PCR Grade sodium salt
Catalog No. 502904983
Encompass
dATP PCR Grade is a 100 mM clear colorless SOLUTIONon of the sodium salt (pH 8.3). PCR Grade nucleotides from Roche are specially manufactured and purified to the highest possible chemical purity. PCR Grade nucleotides contain at least 99% of the relevant deoxynucleoside-triphosphate (dNTP) and less than 0.9% deoxynucleoside-diphosphate (dNDP).
Catalog No. 50-290-4983 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11969013001
dATP PCR Grade is a 100 mM clear colorless SOLUTIONon of the sodium salt (pH 8.3). PCR Grade nucleotides from Roche are specially manufactured and purified to the highest possible chemical purity. PCR Grade nucleotides contain at least 99% of the relevant deoxynucleoside-triphosphate (dNTP) and less than 0.9% deoxynucleoside-diphosphate (dNDP).