Learn More
Revvity Health Sciences Inc dATPαS, [35S]- 1250 Ci/mmol, 12.5 mCi/ml, 1 mCi
Catalog No. 502812315
Encompass
dATPαS, [35S]- 1250 Ci/mmol, 12.5 mCi/ml, 1 mCi
Catalog No. 50-281-2315 Supplier Revvity Health Sciences Inc Supplier No. NEG034H001MC
Add to Cart
Add to Cart
Request a Quote
dATPαS, [35S]- 1250 Ci/mmol, 12.5 mCi/ml, 1 mCi