Learn More
Medchemexpress LLC Dau 5884 hydrochloride | 131780-48-8 | 99.0% | 351.83 g/mol | C17H22ClN3O3 | 10 MG
SDP
Catalog No. 5000242166
Encompass_Preferred
DAU 5884 hydrochloride is the hydrochloride salt of DAU 5884, a potent muscarinic M3 receptor antagonist used in pharmacological research. It inhibits methacholine-dependent effects on cell proliferation and muscle contractility and is supplied as a high-purity reagent for receptor binding and functional studies.
  • Hydrochloride salt form for improved stability and solubility.
  • Potent muscarinic M3 receptor antagonist activity.
  • Inhibits methacholine-dependent cell proliferation and muscle contractility.
  • High purity: 99.0%.
  • Molecular weight 351.83 g/mol; molecular formula C17H22ClN3O3.
  • Supplied in small research quantities suitable for in vitro studies.
Catalog No. 50-002-42166 Supplier Medchemexpress LLC Supplier No. HY10764510MG
DAU 5884 hydrochloride is the hydrochloride salt of DAU 5884, a potent muscarinic M3 receptor antagonist used in pharmacological research. It inhibits methacholine-dependent effects on cell proliferation and muscle contractility and is supplied as a high-purity reagent for receptor binding and functional studies.
  • Hydrochloride salt form for improved stability and solubility.
  • Potent muscarinic M3 receptor antagonist activity.
  • Inhibits methacholine-dependent cell proliferation and muscle contractility.
  • High purity: 99.0%.
  • Molecular weight 351.83 g/mol; molecular formula C17H22ClN3O3.
  • Supplied in small research quantities suitable for in vitro studies.