Learn More
Cayman Chemical DAUDA, 73025-02-2, 25 mg
SDP
Supplier:  Cayman Chemical 1000518825
Encompass_Preferred
A fluorescent fatty acid analog that alters its fluorescent emission spectra and intensity on entry into binding proteins
Catalog No. NC1462592