Learn More
Genscript Corporation Daudi/PD1 Stable Cell Line
Catalog No. 5000059222
Encompass
Recombinant Daudi cells stably overexpress human programmed cell death protein 1 (PD-1) on the surface The surface expression of PD-1 is validated by FACS analysis
Catalog No. 50-000-59222 Supplier Genscript Corporation Supplier No. M00630
Add to Cart
Add to Cart
Request a Quote
Recombinant Daudi cells stably overexpress human programmed cell death protein 1 (PD-1) on the surface The surface expression of PD-1 is validated by FACS analysis