Learn More
Medchemexpress LLC HY-13061 10mg , Daun02 CAS:290304-24-4 Purity:98%
SDP
Catalog No. 501494089
Encompass_Preferred
Medchemexpress, HY-13061 10mg Daun02 CAS:290304-24-4 Formula:C41H44N2O20 Topoisomerase Purity:98% Daun02 is converted by β-galactosidase to Daunorubicin, which is a topoisomerase inhibitor. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-4089 Supplier Medchemexpress LLC Supplier No. HY1306110MG
Medchemexpress, HY-13061 10mg Daun02 CAS:290304-24-4 Formula:C41H44N2O20 Topoisomerase Purity:98% Daun02 is converted by β-galactosidase to Daunorubicin, which is a topoisomerase inhibitor. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.