Learn More
Apexbio Technology LLC Daunorubicin 20830-81-3 10mg
SDP
Catalog No. 5000089116
Encompass_Preferred
Daunorubicin (20830-81-3) is a small-molecule inhibitor targeting DNA topoisomerase II It is designed to inhibit this target thereby disrupting nucleic acid synthesis within tumor cells Daunorubicin exerts its biological activity primarily through stabilizing the topoisomerase II-DNA cleavage complex interfering with DNA replication and transcription In in vitro studies daunorubicin demonstrates inhibition of metabolic incorporation of nucleoside precursors into DNA and RNA with reported IC50 values for inhibition of leukemia cell growth ranging from 0 02 1 M depending on cellular context and assay conditions Based on these pharmacological properties daunorubicin holds research potential in oncology studies related to chemotherapy-induced genotoxicity and nucleic acid metabolism
Catalog No. 50-000-89116 Supplier Apexbio Technology LLC Supplier No. B109910
Daunorubicin (20830-81-3) is a small-molecule inhibitor targeting DNA topoisomerase II It is designed to inhibit this target thereby disrupting nucleic acid synthesis within tumor cells Daunorubicin exerts its biological activity primarily through stabilizing the topoisomerase II-DNA cleavage complex interfering with DNA replication and transcription In in vitro studies daunorubicin demonstrates inhibition of metabolic incorporation of nucleoside precursors into DNA and RNA with reported IC50 values for inhibition of leukemia cell growth ranging from 0 02 1 M depending on cellular context and assay conditions Based on these pharmacological properties daunorubicin holds research potential in oncology studies related to chemotherapy-induced genotoxicity and nucleic acid metabolism