Learn More
Selleck Chemical LLC Daunorubicin HCl 10mM/1mL 23541-50-6 Daunomycin HCl
SDP
Supplier:  Selleck Chemical LLC S303510MM/1ML
Encompass_Preferred
Daunorubicin HCl (Daunomycin) inhibits both DNA and RNA synthesis and inhibits DNA synthesis with Ki of 0.02 ?M in a cell-free assay. Daunorubicin is a topoisomerase II inhibitor that induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0969998