Learn More
Apexbio Technology LLC Daunorubicin HCl 23541-50-6 10mg
SDP
Catalog No. 5000094172
Encompass_Preferred
Daunorubicin HCl (CAS 23541-50-6) is a small-molecule inhibitor targeting DNA topoisomerase II It is designed to inhibit topoisomerase II thereby disrupting nucleic acid synthesis and interfering with DNA replication and transcription Daunorubicin HCl exerts its biological activity primarily through stabilization of DNA-topoisomerase II complexes In in vitro studies daunorubicin exhibits cytotoxic effects with IC50 values typically ranging from 0 1 to 1 M in common leukemia cell assays Based on these pharmacological properties daunorubicin HCl holds research potential in oncology particularly for studying acute myeloid leukemia (AML) and acute lymphoblastic leukemia (ALL)
Catalog No. 50-000-94172 Supplier Apexbio Technology LLC Supplier No. B228510
Daunorubicin HCl (CAS 23541-50-6) is a small-molecule inhibitor targeting DNA topoisomerase II It is designed to inhibit topoisomerase II thereby disrupting nucleic acid synthesis and interfering with DNA replication and transcription Daunorubicin HCl exerts its biological activity primarily through stabilization of DNA-topoisomerase II complexes In in vitro studies daunorubicin exhibits cytotoxic effects with IC50 values typically ranging from 0 1 to 1 M in common leukemia cell assays Based on these pharmacological properties daunorubicin HCl holds research potential in oncology particularly for studying acute myeloid leukemia (AML) and acute lymphoblastic leukemia (ALL)