Learn More
Medchemexpress LLC HY-13062 50mg Medchemexpress, Daunorubicin (Hydrochloride) CAS:23541-50-6 Purity:>98%
SDP
Catalog No. 502028958
Encompass_Preferred
Medchemexpress, HY-13062 50mg Daunorubicin (Hydrochloride) CAS:23541-50-6 Daunorubicin Hydrochloride (Daunomycin Hydrochloride; RP 13057 Hydrochloride; Rubidomycin Hydrochloride) is a topoisomerase II inhibitor with potent antineoplastic activities. Daunorubicin Hydrochloride (Daunomycin Hydrochloride; RP 13057 Hydrochloride; Rubidomycin Hydrochloride) inhibites DNA and RNA synthesis in sensitive and resistant Ehrlich ascites tumor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8958 Supplier Medchemexpress LLC Supplier No. HY1306250MG
Medchemexpress, HY-13062 50mg Daunorubicin (Hydrochloride) CAS:23541-50-6 Daunorubicin Hydrochloride (Daunomycin Hydrochloride; RP 13057 Hydrochloride; Rubidomycin Hydrochloride) is a topoisomerase II inhibitor with potent antineoplastic activities. Daunorubicin Hydrochloride (Daunomycin Hydrochloride; RP 13057 Hydrochloride; Rubidomycin Hydrochloride) inhibites DNA and RNA synthesis in sensitive and resistant Ehrlich ascites tumor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.