Learn More
Medchemexpress LLC Daunorubicin hydrochloride | 23541-50-6 | 99.7% | 500 MG
SDP
Catalog No. 5000339839
Encompass_Preferred
Daunorubicin hydrochloride, also known as Daunomycin, is a potent topoisomerase II inhibitor with significant anti-tumor activity. This anthracycline antibiotic functions by inhibiting DNA and RNA synthesis, leading to the suppression of cancer cell viability. It is recognized for its ability to induce apoptosis and necrosis in target cells.
  • Potent anti-tumor agent
  • Inhibits DNA and RNA synthesis
  • Induces apoptosis and necrosis
  • Anthracycline antibiotic
  • Useful in research for various cancers, including leukemia and lymphomas
Catalog No. 50-003-39839 Supplier Medchemexpress LLC Supplier No. HY13062500MG
Daunorubicin hydrochloride, also known as Daunomycin, is a potent topoisomerase II inhibitor with significant anti-tumor activity. This anthracycline antibiotic functions by inhibiting DNA and RNA synthesis, leading to the suppression of cancer cell viability. It is recognized for its ability to induce apoptosis and necrosis in target cells.
  • Potent anti-tumor agent
  • Inhibits DNA and RNA synthesis
  • Induces apoptosis and necrosis
  • Anthracycline antibiotic
  • Useful in research for various cancers, including leukemia and lymphomas