Learn More
Cayman Chemical Daunorubicin (hydrochloride), Purity: ≥98%, CAS: 23541-50-6, 100 mg
SDP
Catalog No. 502954728
Encompass_Preferred
An antitumor antibiotic that induces apoptosis in mature monocytic U937 and myelocytic HL-60 acute myeloid leukemia cells at 0.2-1 μM; triggers a ROS-dependent sphingomyelin-ceramide pathway that activates the MEKK1-SEK1-JNK
Catalog No. 50-295-4728 Supplier Cayman Chemical Supplier No. 14159100 MG
An antitumor antibiotic that induces apoptosis in mature monocytic U937 and myelocytic HL-60 acute myeloid leukemia cells at 0.2-1 μM; triggers a ROS-dependent sphingomyelin-ceramide pathway that activates the MEKK1-SEK1-JNK