Learn More
Cayman Chemical Daunorubicinone, Purity: ≥98%, CAS: 21794-55-8, 50 mg
SDP
Catalog No. 502948240
Encompass_Preferred
An aglcyone form of the antitumor antibiotic daunorubicin; has been used as a starting material in the synthesis of anticancer agents
Catalog No. 50-294-8240 Supplier Cayman Chemical Supplier No. 3291250 MG
An aglcyone form of the antitumor antibiotic daunorubicin; has been used as a starting material in the synthesis of anticancer agents