Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daunosamine hydrochloride, 85%, MP Biomedicals™

Catalog No. ICN15227325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15227325 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15227325 Supplier MP Biomedicals, Inc Supplier No. 0215227325

Chemical Identifiers

CAS 32385-06-1
Molecular Formula C7H16ClNO3
Molecular Weight (g/mol) 197.66
InChI Key JVYGBUAXDCLPMP-UHFFFAOYNA-N
Synonym daunosamine hydrochloride, methyl l-daunosamine hydrochloride, methyl a-l-daunosamide hcl, methyl, a-l-daunosamine hydrochloride, a:, a approx. 85:15, methyl-a-l-daunosaminide, hydrochloride b-anomer, 2s,3s,4s,6r-4-amino-6-methoxy-2-methyl-tetrahydropyran-3-ol hydrochloride, 2s,3s,4s,6r-4-amino-6-methoxy-2-methyltetrahydro-2h-pyran-3-ol hydrochloride
PubChem CID 55250370

Specifications

Color White
Assay Percent Range 0.85
Quantity 25 mg
Formula Weight 197.7
Physical Form Crystalline Powder
Chemical Name or Material Daunosamine hydrochloride, For TLC analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.