Learn More
Medchemexpress LLC HY-N0220 5mg Medchemexpress, Dauricine CAS:524-17-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN02205MG
Encompass_Preferred
Medchemexpress, HY-N0220 5mg Dauricine CAS:524-17-4 Dauricine, a bisbenzylisoquinoline alkaloid in Asiatic Moonseed Rhizome, possesses anti-inflammatory activity. Dauricine inhibits cell proliferation and invasion, and induces apoptosis by suppressing NF-κB activation in a dose- and time-dependent manner in colon cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2778