Learn More
Medchemexpress LLC dAURK-4 (hydrochloride) | 99.4% | 1071.93 | 100 MG
SDP
Catalog No. 5000351505
Encompass_Preferred
dAURK-4 hydrochloride is a potent and selective PROTAC AURKA (Aurora A) degrader. This derivative of Alisertib exhibits anticancer effects and is composed of a ligand for target protein, a linker, and a ligand for E3 ligase.
  • Potent and selective degrader of AURKA (Aurora A)
  • Exhibits anticancer effects
  • Degrades AURKA in a dose-dependent manner (125-1000 nM; 4-24 hours)
  • Inhibits protein level of AURKA (Aurora A) in MM.1S cells
Catalog No. 50-003-51505 Supplier Medchemexpress LLC Supplier No. HY137344A100MG
dAURK-4 hydrochloride is a potent and selective PROTAC AURKA (Aurora A) degrader. This derivative of Alisertib exhibits anticancer effects and is composed of a ligand for target protein, a linker, and a ligand for E3 ligase.
  • Potent and selective degrader of AURKA (Aurora A)
  • Exhibits anticancer effects
  • Degrades AURKA in a dose-dependent manner (125-1000 nM; 4-24 hours)
  • Inhibits protein level of AURKA (Aurora A) in MM.1S cells