Learn More
Medchemexpress LLC Daurk-4 hydrochloride | 99.4% | 1071.93 | 50 MG
SDP
Catalog No. 5000348520
Encompass_Preferred
dAURK-4 hydrochloride is a potent and selective PROTAC AURKA (Aurora A) degrader, derived from Alisertib. This compound exhibits anticancer effects by targeting specific proteins involved in cell proliferation. It is composed of a ligand for the target protein, a linker, and a ligand for the E3 ligase Thalidomide-O-COOH, enabling dose-dependent degradation of AURKA in vitro.
  • Potent and selective PROTAC AURKA degrader
  • Derivative of Alisertib
  • Exhibits anticancer effects
  • Composed of specific ligands and a linker for targeted degradation
  • Demonstrates dose-dependent degradation of AURKA in vitro
  • High purity (99.44%)
Catalog No. 50-003-48520 Supplier Medchemexpress LLC Supplier No. HY137344A50MG
dAURK-4 hydrochloride is a potent and selective PROTAC AURKA (Aurora A) degrader, derived from Alisertib. This compound exhibits anticancer effects by targeting specific proteins involved in cell proliferation. It is composed of a ligand for the target protein, a linker, and a ligand for the E3 ligase Thalidomide-O-COOH, enabling dose-dependent degradation of AURKA in vitro.
  • Potent and selective PROTAC AURKA degrader
  • Derivative of Alisertib
  • Exhibits anticancer effects
  • Composed of specific ligands and a linker for targeted degradation
  • Demonstrates dose-dependent degradation of AURKA in vitro
  • High purity (99.44%)