Learn More
Cayman Chemical Daursoln 10mg
SDP
Catalog No. 502989302
Encompass_Preferred
An alkaloid with diverse biological activities; enhances camptothecin-induced cytotoxicity in HeLa cells at 10 µM; inhibits camptothecin-induced autophagy in HeLa, A549, and HCT116 cells (IC50s = 74.75, 50.54, and 80.81 μM, respectively); inhibits ADP-induced aggregation of platelets in isolated rabbit whole blood (IC50 = 100 µM); prolongs APD and reduces EADs in papillary muscle preparations isolated from hypertrophied rabbit hearts at 15 µM
Catalog No. 50-298-9302 Supplier Cayman Chemical Supplier No. 2962810 MG
An alkaloid with diverse biological activities; enhances camptothecin-induced cytotoxicity in HeLa cells at 10 µM; inhibits camptothecin-induced autophagy in HeLa, A549, and HCT116 cells (IC50s = 74.75, 50.54, and 80.81 μM, respectively); inhibits ADP-induced aggregation of platelets in isolated rabbit whole blood (IC50 = 100 µM); prolongs APD and reduces EADs in papillary muscle preparations isolated from hypertrophied rabbit hearts at 15 µM