Learn More
Electron Microscopy Sciences Davidson's (Hartmann's) Fixative 4 LT
SDP
Catalog No. NC1120776
Encompass_Preferred
This is rapid fixative. Results with good nuclear detail and minimal formalin pigment. Hatmann's (Davidson's) Fixative is useful for preparing tumors, bone marrow, gynecologic specimen, fatty breast, medical biopsies. Fixation of small specimen is rapid. Exposure to the fixative should be limited to 24 hours (tissue may then be transferred to neutral buffered formalin or 70% alcohol for storage).
Based on the volume of 1125 ml solution, there is 2 part Formalin (35-39%), 3 part Ethanol, 1 part Glacial Acetic acid, and 3 part of tap water.
Catalog No. NC1120776 Supplier Electron Microscopy Sciences Supplier No. 6413314
This is rapid fixative. Results with good nuclear detail and minimal formalin pigment. Hatmann's (Davidson's) Fixative is useful for preparing tumors, bone marrow, gynecologic specimen, fatty breast, medical biopsies. Fixation of small specimen is rapid. Exposure to the fixative should be limited to 24 hours (tissue may then be transferred to neutral buffered formalin or 70% alcohol for storage).
Based on the volume of 1125 ml solution, there is 2 part Formalin (35-39%), 3 part Ethanol, 1 part Glacial Acetic acid, and 3 part of tap water.