Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Davidson's Fixative Modified Yellow
SDP

Catalog No. 21402129
Change view
Click to view available options
Solution Type:
Formaldehyde
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Solution Type
21-402-129 Formaldehyde
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-402-129 Supplier StatLab Supplier No. 0231ALLE01YL
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Modified fixative Yellow, routinely used for optimal fixation of eyes

  • Davidson fixed eyes were easier to cut because it had a more robust structure compared to the formalin fixed eyes
  • This fixative can support excellent staining of acrosomes with Periodic Acid-Schiff (PAS) reagent when staging of spermatogenesis is required

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Solution Type Formaldehyde
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.