Learn More
Sigma Aldrich Fine Chemicals Biosciences Davis Minimal Broth | 500g
Catalog No. NC1746865
Encompass
Minimal medium recommended for the isolation and characterization of nutritional mutants of Escherichia coli.
Catalog No. NC1746865 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 15758500GF
Minimal medium recommended for the isolation and characterization of nutritional mutants of Escherichia coli.