Learn More
Stoelting™ Davis Retractor
SDP
Catalog No. 10001422 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
3 x 3 blunt prongs
Catalog No. Model
10-001-422 3 x 3 blunt prongs
1 options
Catalog No. 10-001-422 Supplier Stoelting™ Supplier No. 5212621
Add to Cart
Add to Cart
Stoelting™ Davis Retractors can be used to actively separate the edges of a surgical incision or wound or hold back underlying organs and tissues so that body parts under the incision may be accessed.
  • Sharp retractors are better at providing a secure grip and they can safely hold back the skin, muscles, fascia, tendons, or the periosteum but they may damage vessels, nerves, the peritoneum, glands or cysts
  • Blunt retractors are recommended when iatrogenic injury is a concern.
  • The main benefit of blunt retractors is that they prevent unintended injury to the tissue held by the retractor or to the underlying tissue.
Specifications
For Use With (Application) Neuroscience
Length (English) 4.5 in.
Model 3 x 3 blunt prongs