Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dukal™ DawnMist™ Denture Tablets

Catalog No. 22415780
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-415-780 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-415-780 Supplier Dukal™ Supplier No. DEN6290
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These Denture Tablets are antibacterial, remove stains and control odors.

  • Antibacterial formula with baking soda.
  • Helps to remove stains and control odors.
  • Individually foil wrapped.
  • Made in the USA.
  • Compared to Fixodent.

Specifications

Product Type Denture Tablets
Color White
Material Sodium Bicarbonate, Citric Acid, Sodium Carbonate, Sodium Percarbonate, Magnesium Stearete
For Use With (Application) Denture Cleansing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.