Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dukal™ DawnMist™ Hair Conditioner

Catalog No. 22415787
Change view
Click to view available options
Quantity:
2 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-415-787 2 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-415-787 Supplier Dukal™ Supplier No. HC02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Hair Conditioner moisturizes hair, leaving it soft and manageable.

  • Moisturizes hair and scalp
  • Easy rinse formula
  • Leaves hair soft and manageable
  • Alcohol free

Specifications

Product Type Hair Conditioner
Color Ivory
Composition Alcohol Free Liquid
For Use With (Application) Hair Care
Quantity 2 oz.
Volume (English) 2 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.