Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dukal™ DawnMist™ Rinse Free Shampoo and Body Bath

Catalog No. 22415789
Change view
Click to view available options
Quantity:
2 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-415-789 2 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-415-789 Supplier Dukal™ Supplier No. NRB4463
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Rinse Free Shampoo and Body Bath is a mild, pH balanced formula that helps maintain skin moisture.

  • Cleanses without the need for water
  • Mild
  • pH balanced formula helps maintain skins moisture

Specifications

Product Type Shampoo and Body Bath
Color Clear
Composition Liquid pH Balanced Formula
For Use With (Application) Personal Cleansing
Quantity 2 oz.
Volume (English) 2 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.