Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dukal™ DawnMist™ Shampoo Caps

Catalog No. 22415796
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-415-796 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-415-796 Supplier Dukal™ Supplier No. SC3756
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These Rinse Free Shampoo Caps feature an all-in-one hair cleansing system that requires no water.

  • A convenient all-in-one hair cleansing system.
  • Cleanses and conditions hair without the need for a sink or water.
  • Soothing
  • Sanitary
  • pH balanced

Specifications

Product Type Cleansing Cap
Color White
Composition Liquid Filled Cap
For Use With (Application) Affix to the head
For Use With (Equipment) Hair Cleansing
Weight (English) 1 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.