Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dukal™ DawnMist™ Tearless Baby Shampoo

Catalog No. 22415800
Change view
Click to view available options
Quantity:
2 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-415-800 2 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-415-800 Supplier Dukal™ Supplier No. TS4470
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Tearless Baby Shampoo is a pH balanced formula that is great for babies and adults.

  • pH balanced formula gentle on hair, scalp, and eyes.
  • Leaves hair soft and manageable.
  • Light baby fresh scent.
  • Great for babies and adults.

Specifications

Product Type Shampoo
Color Orange
Composition Liquid
For Use With (Application) Hair Cleansing
Quantity 2 oz.
Volume (English) 2 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.