Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dukal™ DawnMist™ Toothbrushes

Catalog No. 22415798
Change view
Click to view available options
No. of Tufts:
30 Tufts
52 Tufts
Color:
Blue with White Bristles
Ivory with Clear Bristles
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tufts Color
22-415-798 30 Tufts Ivory with Clear Bristles
22-415-799 52 Tufts Blue with White Bristles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-415-798 Supplier Dukal™ Supplier No. TB30
Add to Cart
Add to Cart
This item is not returnable. View return policy

These Toothbrushes feature soft bristles with rounded polish tips.

  • Features soft bristles with rounded polish tips.
  • Individually packaged.

Specifications

Product Type Toothbrush
Color Ivory with Clear Bristles
Material Plastic
For Use With (Application) Oral Care
For Use With (Equipment) Toothpaste
No. of Tufts 30 Tufts
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.