Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dukal™ DawnMist™ Toothpaste

Catalog No. 22415794
Change view
Click to view available options
Volume (English):
0.6 oz.
0.85 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
22-415-794 0.85 oz.
22-415-793 0.6 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-415-794 Supplier Dukal™ Supplier No. RTP085
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Toothpaste contains cavity fighting fluoride and is designed for sensitive users.

  • Contains cavity fighting fluoride.
  • Pleasant mint taste leaves mouth feeling fresh and clean.
  • Gentle for sensitive users.

Specifications

Product Type Toothpaste
Color White
Material Fluoride Paste
For Use With (Application) Oral Care
For Use With (Equipment) Toothbrush
Volume (English) 0.85 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.