Learn More
Cayman Chemical DAX-1NROB1 Monclonal ntI 100uL
SDP
Catalog No. 502948387
Encompass_Preferred
An active metabolite of thioridazine formed via sulfoxidation of thioridazine by CYP2D6 in human liver microsomes binds to histamine H1 dopamine D2 muscarinic acetylcholine and a1- and a2-adrenergic receptors (Kds 1.8 19 69 2 and 1600 nM respectively) prevents mescaline-induced hyperactivity in mice at 10 30 and 45 mg/kg
Catalog No. 50-294-8387 Supplier Cayman Chemical Supplier No. 32763100 UL
An active metabolite of thioridazine formed via sulfoxidation of thioridazine by CYP2D6 in human liver microsomes binds to histamine H1 dopamine D2 muscarinic acetylcholine and a1- and a2-adrenergic receptors (Kds 1.8 19 69 2 and 1600 nM respectively) prevents mescaline-induced hyperactivity in mice at 10 30 and 45 mg/kg