Learn More
Cayman Chemical DAX-1NROB1 Monclonal ntI 100uL
SDP
Supplier:  Cayman Chemical 32764100 UL
Encompass_Preferred
An active metabolite of thioridazine formed via sulfoxidation of thioridazine by CYP2D6 in human liver microsomes binds to histamine H1 dopamine D2 muscarinic acetylcholine and a1- and a2-adrenergic receptors (Kds 1.8 19 69 2 and 1600 nM respectively) prevents mescaline-induced hyperactivity in mice at 10 30 and 45 mg/kg
Catalog No. 50-294-7539