Learn More
Enzo Life Sciences Daxx (human) monoclonal antibody (DAXX-01) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804404C100
Encompass_Preferred
Clone: DAXX-01. Isotype: IgG1. Immunogen: Recombinant human Daxx (aa 558-740). Species reactivity: Human. Applications: ICC, WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 15mM sodium azide.
Catalog No. 50-200-9903