Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Humboldt Day Pinchcock
SDP

Catalog No. 05867
Change view
Click to view available options
Length (Metric):
6.65 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
05-867 6.65 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-867 Supplier Humboldt Supplier No. H8630
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Can be put on or removed without disconnecting apparatus

Can be put on or removed without disconnecting apparatus

  • Tough 302 Stainless Steel
  • Holds flexible tubing up to 0.37 in. (9.5mm) O.D.

Specifications

Quantity 1 Ea.
Type Pinchcock Clamp
Material Stainless Steel (302)
Length (English) 2.62 in.
Length (Metric) 6.65 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.