Learn More
Aviva Systems Biology Corp DAZ2 Antibody - N-terminal region (ARP40250_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40250_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DAZ2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human DAZ2. Alternate names: pDP1678ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53181