Learn More
Aviva Systems Biology Corp DAZ4 Antibody - middle region (ARP40242_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40242_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DAZ4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DAZ4. Alternate names: DAZ, DAZ1, pDP1680, pDP1681ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57363