Learn More
Aviva Systems Biology Corp DAZ4 Antibody - N-terminal region (ARP40243_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40243_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DAZ4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human DAZ4. Alternate names: pDP1680, pDP1681ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53180