Learn More
Aviva Systems Biology Corp DAZAP1 Antibody - C-terminal region (ARP40927_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40927_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DAZAP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human DAZAP1. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-57666