Learn More
Aviva Systems Biology Corp DAZAP1 Antibody - C-terminal region (ARP41161_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41161_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DAZAP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human DAZAP1. Alternate names: MGC19907ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-53284