Learn More
TARGETMOL CHEMICALS INC Dazostinag disodium 1MG
SDP
Catalog No. 502878217
Encompass_Preferred
Also available in 5 mg, 25 mg and bulk. Please contact Fisher for quotes. Dazostinag disodium (TAK-676) is a synthetic novel interferon gene (STING) agonist that triggers STING signaling pathway activation and type I interferon activation. Dazostinag disodium (TAK-676) is also a highly effective immune system modulator with complete resolution and lasting memory of T cell immunity and the ability to promote lasting interferon-dependent anti-tumor immune responses. Purity 99.96%
Catalog No. 50-287-8217 Supplier TARGETMOL CHEMICALS INC Supplier No. T724821MG
Also available in 5 mg, 25 mg and bulk. Please contact Fisher for quotes. Dazostinag disodium (TAK-676) is a synthetic novel interferon gene (STING) agonist that triggers STING signaling pathway activation and type I interferon activation. Dazostinag disodium (TAK-676) is also a highly effective immune system modulator with complete resolution and lasting memory of T cell immunity and the ability to promote lasting interferon-dependent anti-tumor immune responses. Purity 99.96%