Learn More
Medchemexpress LLC Dazostinag disodium | 2553413-93-5 | 98.2% | 754.48 g/mol | C21H20F2N8Na2O10P2S2 | 10 MG
SDP
Catalog No. 5000287083
Encompass_Preferred
Dazostinag disodium is the disodium salt form of a STING agonist supplied for preclinical research to activate STING signaling and induce type I interferon responses. The material is provided as a high-purity, water-soluble solid for use in immunology and oncology studies; supporting documents (data sheet, COA, SDS) describe analytical data, handling, and storage recommendations.
  • Disodium salt form for improved aqueous solubility.
  • High purity suitable for preclinical research.
  • White to off-white solid appearance.
  • Soluble in water (≥ 100 mg/mL) for in vitro applications.
  • Available in small research package sizes for dosing studies.
  • Store sealed at -20°C, avoid moisture; in solution keep at -80°C for long term.
Catalog No. 50-002-87083 Supplier Medchemexpress LLC Supplier No. HY14802910MG
Add to Cart
Add to Cart
Request a Quote
Dazostinag disodium is the disodium salt form of a STING agonist supplied for preclinical research to activate STING signaling and induce type I interferon responses. The material is provided as a high-purity, water-soluble solid for use in immunology and oncology studies; supporting documents (data sheet, COA, SDS) describe analytical data, handling, and storage recommendations.
  • Disodium salt form for improved aqueous solubility.
  • High purity suitable for preclinical research.
  • White to off-white solid appearance.
  • Soluble in water (≥ 100 mg/mL) for in vitro applications.
  • Available in small research package sizes for dosing studies.
  • Store sealed at -20°C, avoid moisture; in solution keep at -80°C for long term.