Learn More
Medchemexpress LLC Dazostinag disodium | 2553413-93-5 | 98.7% | 754.48 | 500 UG
SDP
Catalog No. 5000359747
Encompass_Preferred
Dazostinag disodium (TAK-676) is a STING agonist that activates the STING signaling pathway and type I interferons. It acts as a modulator of the immune system, leading to complete regressions and durable memory T-cell immunity, and promotes durable IFN-dependent antitumor immunity.
  • Activates the STING-TBK1-IRF3 pathway
  • Exerts in vitro immune cell activation in mouse BM-derived dendritic cells
  • Promotes activation of dendritic cells, natural killer cells, and T cells
  • Well tolerated and shows dose-proportional pharmacokinetics in plasma
  • Exhibits anti-tumor function in syngeneic tumors in mice
  • Induces dose-dependent cytokine responses, increasing activation and proliferation of immune cells
Catalog No. 50-003-59747 Supplier Medchemexpress LLC Supplier No. HY148029500UG
Dazostinag disodium (TAK-676) is a STING agonist that activates the STING signaling pathway and type I interferons. It acts as a modulator of the immune system, leading to complete regressions and durable memory T-cell immunity, and promotes durable IFN-dependent antitumor immunity.
  • Activates the STING-TBK1-IRF3 pathway
  • Exerts in vitro immune cell activation in mouse BM-derived dendritic cells
  • Promotes activation of dendritic cells, natural killer cells, and T cells
  • Well tolerated and shows dose-proportional pharmacokinetics in plasma
  • Exhibits anti-tumor function in syngeneic tumors in mice
  • Induces dose-dependent cytokine responses, increasing activation and proliferation of immune cells